COM448687866 Combi-Blocks, Inc.

4-(Methanesulfonylmethyl)-n-propylbenzamide

  • MDL number MFCD15343016
  • Cas Number : 1182050-90-3
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0045

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0045
Availability details for
Quantity
  • Availability
  • Add to Cart