COM448687868 Combi-Blocks, Inc.

2-[2-Fluoro-3-(methanesulfonylmethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD18758211
  • Molecular Weight : 314.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0047

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0047
Availability details for
Quantity
  • Availability
  • Add to Cart