COM448687871 Combi-Blocks, Inc.

1-[4-(Methanesulfonylmethyl)benzoyl]pyrrolidine

  • MDL number MFCD13511196
  • Cas Number : 1209244-87-0
  • Molecular Weight : 267.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0050

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0050
Availability details for
Quantity
  • Availability
  • Add to Cart