COM448687880 Combi-Blocks, Inc.

1-[4-(Methanesulfonylmethyl)phenyl]ethanone

  • MDL number MFCD26734679
  • Cas Number : 1309803-50-6
  • Molecular Weight : 212.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0059

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0059
Availability details for
Quantity
  • Availability
  • Add to Cart