COM448687881 Combi-Blocks, Inc.

4-Methanesulfonyl-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD18633175
  • Cas Number : 1820664-84-3
  • Molecular Weight : 223.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0060

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0060
Availability details for
Quantity
  • Availability
  • Add to Cart