COM448687883 Combi-Blocks, Inc.

3-(1-Methanesulfonylethyl)aniline

  • MDL number MFCD20724197
  • Cas Number : 773846-47-2
  • Molecular Weight : 199.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0062

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0062
Availability details for
Quantity
  • Availability
  • Add to Cart