COM448687885 Combi-Blocks, Inc.

2-Bromo-5-(methanesulfonylmethyl)aniline

  • MDL number MFCD24229901
  • Cas Number : 1545654-97-4
  • Molecular Weight : 264.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0064

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0064
Availability details for
Quantity
  • Availability
  • Add to Cart