COM448687888 Combi-Blocks, Inc.

4-Methyl-3-(methysulfonylmethyl)phenylboronic acid pinacol ester

  • MDL number MFCD18758220
  • Cas Number : 2377611-56-6
  • Molecular Weight : 310.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0067
Availability details for
Quantity
  • Availability
  • Add to Cart