COM448687891 Combi-Blocks, Inc.

4-Chloro-2-(methanesulfonylmethyl)aniline

  • MDL number MFCD21957720
  • Cas Number : 1101063-19-7
  • Molecular Weight : 219.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0070

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0070
Availability details for
Quantity
  • Availability
  • Add to Cart