COM456878633 Combi-Blocks, Inc.

4-(Benzyloxy)-5-bromobenzene-1,2-diamine hydrochloride

  • MDL number MFCD22192365
  • Cas Number : 1951439-28-3
  • Molecular Weight : 329.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3708

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3708
Availability details for
Quantity
  • Availability
  • Add to Cart