COM456878634 Combi-Blocks, Inc.

N-Methyl 4-Acetamido-2-methylbenzenesulfonamide

  • MDL number MFCD28962744
  • Cas Number : 1845715-32-3
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6072

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6072
Availability details for
Quantity
  • Availability
  • Add to Cart