COM456878635 Combi-Blocks, Inc.

4-[(1,3-Dioxoisoindol-2-yl)methyl]-N-(2-nitrophenyl)benzamide

  • MDL number MFCD28962745
  • Cas Number : 1845706-31-1
  • Molecular Weight : 401.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6076

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6076
Availability details for
Quantity
  • Availability
  • Add to Cart