COM456878636 Combi-Blocks, Inc.

4-methyl-3-(trifluoromethyl)aniline hydrochloride

  • MDL number MFCD28962747
  • Cas Number : 1845696-01-6
  • Molecular Weight : 211.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6083

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6083
Availability details for
Quantity
  • Availability
  • Add to Cart