COM456878637 Combi-Blocks, Inc.

N-[2-Nitro-5-(trifluoromethyl)phenyl]methanesulfonamide

  • MDL number MFCD28962748
  • Cas Number : 1845706-41-3
  • Molecular Weight : 284.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6087

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6087
Availability details for
Quantity
  • Availability
  • Add to Cart