COM456878639 Combi-Blocks, Inc.

4-(Methoxycarbonyl)-2-(trifluoromethyl)phenylboronic acid

  • MDL number MFCD28515374
  • Molecular Weight : 248.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-1718

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-1718
Availability details for
Quantity
  • Availability
  • Add to Cart