COM456878640 Combi-Blocks, Inc.

3-Bromo-2-fluoro-6-methylphenylboronic acid

  • MDL number MFCD28515375
  • Cas Number : 2246650-88-2
  • Molecular Weight : 232.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3575

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3575
Availability details for
Quantity
  • Availability
  • Add to Cart