COM456878641 Combi-Blocks, Inc.

[5-Chloro-2-(morpholin-4-yl)phenyl]boronic acid

  • MDL number MFCD28962749
  • Cas Number : 2377605-70-2
  • Molecular Weight : 241.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4635

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4635
Availability details for
Quantity
  • Availability
  • Add to Cart