COM456878642 Combi-Blocks, Inc.

(2-{4-[(tert-Butoxy)carbonyl]piperazin-1-yl}-4-chlorophenyl)boronic acid

  • MDL number MFCD28515376
  • Cas Number : 2377606-54-5
  • Molecular Weight : 340.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4862
Availability details for
Quantity
  • Availability
  • Add to Cart