COM456878644 Combi-Blocks, Inc.

(4-{[2-(Trimethylsilyl)ethoxy]carbonyl}phenyl)boronic acid

  • MDL number MFCD28515378
  • Molecular Weight : 266.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1081

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1081
Availability details for
Quantity
  • Availability
  • Add to Cart