COM456878646 Combi-Blocks, Inc.

3-Bromo-2-fluoro-4-methylphenylboronic acid

  • MDL number MFCD28515380
  • Molecular Weight : 232.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1102
Availability details for
Quantity
  • Availability
  • Add to Cart