COM456878647 Combi-Blocks, Inc.

2-(Benzyloxy)-5-cyanophenylboronic acid

  • MDL number MFCD28515381
  • Molecular Weight : 253.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1104

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1104
Availability details for
Quantity
  • Availability
  • Add to Cart