COM456878648 Combi-Blocks, Inc.

5-(BOC-Aminomethyl)thiophene-3-boronic acid

  • MDL number MFCD28515382
  • Cas Number : 2246751-62-0
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1107

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1107
Availability details for
Quantity
  • Availability
  • Add to Cart