COM456878651 Combi-Blocks, Inc.

1-(tert-Butoxycarbonyl)-3-isopropylpyrazole-4-boronic acid

  • MDL number MFCD28515385
  • Cas Number : 2304635-44-5
  • Molecular Weight : 254.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1111

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1111
Availability details for
Quantity
  • Availability
  • Add to Cart