COM456878655 Combi-Blocks, Inc.

[3-({[(tert-Butoxy)carbonyl]amino}methyl)-5-fluorophenyl]boronic acid

  • MDL number MFCD28962752
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1139

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1139
Availability details for
Quantity
  • Availability
  • Add to Cart