COM456878657 Combi-Blocks, Inc.

3-(N-Boc-N-methylamino)methylphenylboronic acid

  • MDL number MFCD28962754
  • Cas Number : 2246563-06-2
  • Molecular Weight : 265.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1147

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1147
Availability details for
Quantity
  • Availability
  • Add to Cart