COM456878658 Combi-Blocks, Inc.

2,3-Dichloro-5-methylphenylboronic acid

  • MDL number MFCD28787023
  • Cas Number : 2377606-78-3
  • Molecular Weight : 204.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1148

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1148
Availability details for
Quantity
  • Availability
  • Add to Cart