COM456878660 Combi-Blocks, Inc.

(2E)-4,4-Dimethoxy-3-methylbut-2-enenitrile

  • MDL number MFCD28515387
  • Cas Number : 84586-19-6
  • Molecular Weight : 141.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5824

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5824
Availability details for
Quantity
  • Availability
  • Add to Cart