COM456878662 Combi-Blocks, Inc.

2-Fluoro-6-methoxy-3-methylphenylboronic acid pinacol ester

  • MDL number MFCD28962757
  • Molecular Weight : 266.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2130

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2130
Availability details for
Quantity
  • Availability
  • Add to Cart