COM456878665 Combi-Blocks, Inc.

5-(Furan-2-yl)-2H-pyrazol-3-amine hydrochloride

  • MDL number MFCD28962758
  • Molecular Weight : 185.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4269

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4269
Availability details for
Quantity
  • Availability
  • Add to Cart