COM456878666 Combi-Blocks, Inc.

3-Bromo-2-fluoro-6-methylphenol

  • MDL number MFCD28739775
  • Cas Number : 1807088-23-8
  • Molecular Weight : 205.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4287

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4287
Availability details for
Quantity
  • Availability
  • Add to Cart