COM456878667 Combi-Blocks, Inc.

4-Bromo-1-isopentyl-3-(trifluoromethyl)pyrazole

  • MDL number MFCD28962759
  • Cas Number : 1845690-56-3
  • Molecular Weight : 285.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2233

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2233
Availability details for
Quantity
  • Availability
  • Add to Cart