COM456878668 Combi-Blocks, Inc.

6-Nitro-1,2,3a,7a-tetrahydroindazol-3-one

  • MDL number MFCD28962760
  • Cas Number : 1845716-90-6
  • Molecular Weight : 181.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0530

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?IN-0530
Availability details for
Quantity
  • Availability
  • Add to Cart