COM456878669 Combi-Blocks, Inc.

N-(3-methoxypropyl)-N-(2-nitrophenyl)methanesulfonamide

  • MDL number MFCD28515391
  • Cas Number : 1845698-50-1
  • Molecular Weight : 288.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1228

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1228
Availability details for
Quantity
  • Availability
  • Add to Cart