COM456878671 Combi-Blocks, Inc.

N-(2-Methoxyethyl)-N-(2-nitrophenyl)methanesulfonamide

  • MDL number MFCD28515393
  • Cas Number : 1845706-34-4
  • Molecular Weight : 274.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1232

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1232
Availability details for
Quantity
  • Availability
  • Add to Cart