COM456878672 Combi-Blocks, Inc.

2-Methanesulfonyl-5-nitrobenzonitrile

  • MDL number MFCD28962761
  • Cas Number : 180297-57-8
  • Molecular Weight : 226.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1233

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1233
Availability details for
Quantity
  • Availability
  • Add to Cart