COM456878674 Combi-Blocks, Inc.

4-Bromo-5-(tributylstannyl)-1,3-thiazole

  • MDL number MFCD28515395
  • Molecular Weight : 453.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-7991

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OS-7991
Availability details for
Quantity
  • Availability
  • Add to Cart