COM456878690 Combi-Blocks, Inc.

tert-Butyl N-[3-isopropyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate

  • MDL number MFCD28962770
  • Cas Number : 2377606-19-2
  • Molecular Weight : 361.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1144

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1144
Availability details for
Quantity
  • Availability
  • Add to Cart