COM456878694 Combi-Blocks, Inc.

4-(3-Methylbutanoyl)phenylboronic acid pinacol ester

  • MDL number MFCD28515405
  • Cas Number : 2377611-71-5
  • Molecular Weight : 288.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6425

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6425
Availability details for
Quantity
  • Availability
  • Add to Cart