COM456878698 Combi-Blocks, Inc.

3-Bromo-6-chloro-N-methylpyridin-2-amine

  • MDL number MFCD28515408
  • Cas Number : 1845690-58-5
  • Molecular Weight : 221.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2080

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2080
Availability details for
Quantity
  • Availability
  • Add to Cart