COM456878699 Combi-Blocks, Inc.

2-Chloro-6-(methylamino)pyridine-3-carbonitrile

  • MDL number MFCD28515409
  • Cas Number : 1845689-90-8
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2081

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2081
Availability details for
Quantity
  • Availability
  • Add to Cart