COM456878700 Combi-Blocks, Inc.

tert-butyl N-(5-bromo-6-chloropyridin-2-yl)carbamate

  • MDL number MFCD28515410
  • Cas Number : 1845713-77-0
  • Molecular Weight : 307.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2082

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2082
Availability details for
Quantity
  • Availability
  • Add to Cart