COM456878701 Combi-Blocks, Inc.

5,6-Dihdyro-6,6-dimethylindolo[1,2-a]quinoxaline HCl

  • MDL number MFCD28515411
  • Cas Number : 1845716-92-8
  • Molecular Weight : 284.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2090
Availability details for
Quantity
  • Availability
  • Add to Cart