COM456878702 Combi-Blocks, Inc.

Methyl 3,5-dibromo-6-hydroxypyridine-2-carboxylate

  • MDL number MFCD28515412
  • Cas Number : 1845690-59-6
  • Molecular Weight : 310.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2098

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2098
Availability details for
Quantity
  • Availability
  • Add to Cart