COM456878703 Combi-Blocks, Inc.

5-Fluoro-2,6-dimethoxypyridine-3-carbonitrile

  • MDL number MFCD28962772
  • Cas Number : 1845696-07-2
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2100

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2100
Availability details for
Quantity
  • Availability
  • Add to Cart