COM456878705 Combi-Blocks, Inc.

N-[(2R)-2-Amino-2-phenylethyl]aniline

  • MDL number MFCD28515413
  • Cas Number : 1501975-72-9
  • Molecular Weight : 212.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6004

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6004
Availability details for
Quantity
  • Availability
  • Add to Cart