COM456878706 Combi-Blocks, Inc.

tert-Butyl 4-acetyl-4-methylpiperidine-1-carboxylate

  • MDL number MFCD28962773
  • Cas Number : 1507372-37-3
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 97
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6030
Availability details for
Quantity
  • Availability
  • Add to Cart