COM456878710 Combi-Blocks, Inc.

Ethyl 4-(butylamino)-3-nitrobenzoate

  • MDL number MFCD28962778
  • Cas Number : 1206724-39-1
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9323

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9323
Availability details for
Quantity
  • Availability
  • Add to Cart