COM456878712 Combi-Blocks, Inc.

Ethyl 1-butyl-2-methyl-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD28962780
  • Cas Number : 1845689-91-9
  • Molecular Weight : 260.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9326
Availability details for
Quantity
  • Availability
  • Add to Cart