COM456878713 Combi-Blocks, Inc.

Ethyl 1-butyl-1,2,3-benzotriazole-5-carboxylate

  • MDL number MFCD28962781
  • Cas Number : 1845698-42-1
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9327

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9327
Availability details for
Quantity
  • Availability
  • Add to Cart