COM456878714 Combi-Blocks, Inc.

Ethyl 1-tert-butyl-1,2,3-benzotriazole-5-carboxylate

  • MDL number MFCD28962782
  • Cas Number : 1845716-98-4
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9332

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?WZ-9332
Availability details for
Quantity
  • Availability
  • Add to Cart